5-Norbornene-2-endo,3-endo-dimethanol ≥98.0%

Supplier: TCI America

Synonyms: Bicyclo[2.2.1]hept-5-ene-2-endo,3-endo-dimethanol, 5-endo,6-endo-Bis(hydroxymethyl)-2-norbornene

N0768-5G N0768-1G
TCN0768-5GEA 373.56 CAD
TCN0768-5G TCN0768-1G
5-Norbornene-2-endo,3-endo-dimethanol ≥98.0%
5-Norbornene-2-endo,3-endo-dimethanol

CAS Number: 699-97-8
Molecular Formula: C9H14O2
Molecular Weight: 154.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 86


Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2-endo,3-endo-dimethanol, 5-endo,6-endo-Bis(hydroxymethyl)-2-norbornene

Formula: C₉H₁₄O₂
MW: 154.21 g/mol
Boiling Pt: 130 °C
Melting Pt: 86 °C
MDL Number: MFCD00674044
CAS Number: 699-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8074bb6f505c714386e96736060e36a447f7fdf1&vid=8074bb6f505c714386e96736060e36a447f7fdf1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR