cis-5-Norbornene exo-2,3-dicarboxylic anhydride ≥97.0%
Supplier: TCI America
Synonyms:
(3aR,4R,7S,7aS)-rel-3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione, Bicyclo[2.2.1]hept-5-ene-exo-2,3-dicarboxylic Anhydride
N0767-1G
N0767-5G
TCN0767-1GEA
63.2
CAD
TCN0767-1G
TCN0767-5G
cis-5-Norbornene exo-2,3-dicarboxylic anhydride ≥97.0%
cis-5-Norbornene exo-2,3-dicarboxylic anhydride
CAS Number: 2746-19-2
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 144
Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-exo-2,3-dicarboxylic Anhydride
Formula:
C₉H₈O₃ MW: 164.16 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD00630580 CAS Number: 2746-19-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...