1,8-Naphthosultone ≥98.0% (by HPLC)

Supplier: TCI America

N0766-25G N0766-5G
TCN0766-25GEA 406.06 CAD
TCN0766-25G TCN0766-5G
1,8-Naphthosultone ≥98.0% (by HPLC)
1,8-Naphthosultone

CAS Number: 83-31-8
Molecular Formula: C10H6O3S
Molecular Weight: 206.22
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 158


Other Product Names / Synonyms: 8-Hydroxynaphthalene-1-sulfonic Acid Sultone, 1,8-Naphthalenesultone, 1-Naphthol-8-sulfonic Acid Sultone

Formula: C₁₀H₆O₃S
Melting Pt: 158 °C
CAS Number: 83-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dbbc755f0002e31e9ca28d63b732ff5aadea78a&vid=8dbbc755f0002e31e9ca28d63b732ff5aadea78a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR