5-Norbornene-2,3-dimethanol ≥95.0%

Supplier: TCI America

N0764-1G N0764-5G N0764-25G
TCN0764-1GEA 167.48 CAD
TCN0764-1G TCN0764-5G TCN0764-25G
5-Norbornene-2,3-dimethanol ≥95.0%
5-Norbornene-2,3-dimethanol

CAS Number: 85-39-2
Molecular Formula: C9H14O2
Molecular Weight: 154.21
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 82


Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2,3-dimethanol, 2,3-Bis(hydroxymethyl)-5-norbornene

Formula: C₉H₁₄O₂
Melting Pt: 82 °C
MDL Number: MFCD00474435
CAS Number: 85-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3471bbecac85c6f8c212fc367d5b167a729ac05&vid=c3471bbecac85c6f8c212fc367d5b167a729ac05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR