N-(4-Oxocyclohexyl)acetamide ≥98.0%
Supplier: TCI America
A1929-25G
A1929-5G
TCA1929-25GEA
71.1
CAD
TCA1929-25G
TCA1929-5G
N-(4-Oxocyclohexyl)acetamide ≥98.0%
N-(4-Oxocyclohexyl)acetamide
CAS Number: 27514-08-5
MDL Number: MFCD03703462
Molecular Formula: C8H13NO2
Molecular Weight: 155.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 137
Other Product Names / Synonyms: N-(4-Oxocyclohexyl)acetamide
Formula:
C₈H₁₃NO₂ Melting Pt: 137 °C |
MDL Number:
MFCD03703462 CAS Number: 27514-08-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...