2,3-Norbornanedicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Bicyclo[2.2.1]heptane-2,3-dicarboxylic Acid
N0753-5G
N0753-25G
TCN0753-5GEA
168.59
CAD
TCN0753-5G
TCN0753-25G
2,3-Norbornanedicarboxylic acid ≥98.0% (by GC, titration analysis)
2,3-Norbornanedicarboxylic acid
CAS Number: 1724-08-9
Molecular Formula: C9H12O4
Molecular Weight: 184.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bicyclo[2.2.1]heptane-2,3-dicarboxylic Acid
Formula:
C₉H₁₂O₄ MW: 184.19 g/mol |
MDL Number:
MFCD00520577 CAS Number: 1724-08-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...