Potassium bisnonafluoro-1-butanesulfonimidate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
N0712-1G
N0712-5G
TCN0712-1GEA
154.84
CAD
TCN0712-1G
TCN0712-5G
Potassium bisnonafluoro-1-butanesulfonimidate ≥98.0% (by titrimetric analysis)
Potassium bisnonafluoro-1-butanesulfonimidate
CAS Number: 129135-87-1
MDL Number: MFCD06200794
Molecular Formula: C8HF18NO4S2
Molecular Weight: 619.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Flash Point (°C): 40
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: Bisnonafluoro-1-butanesulfonimide Potassium Salt, Bis(nonafluoro-1-butanesulfonyl)amine Potassium Salt, Nonafluoro-1-butanesulfonimide Potassium Salt, Potassium Nonafluoro-1-butanesulfonimidate
Formula:
C₈HF₁₈NO₄S₂ Flash Pt: 40 °C |
MDL Number:
MFCD06200794 CAS Number: 129135-87-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...