4-Amylcyclohexanol (cis- and trans- mixture) ≥98.0%

Supplier: TCI America

A1926-25G A1926-5G
TCA1926-25GEA 434.66 CAD
TCA1926-25G TCA1926-5G
4-Amylcyclohexanol (cis- and trans- mixture) ≥98.0%
4-Amylcyclohexanol (cis- and trans- mixture)

CAS Number: 54410-90-1
Molecular Formula: C11H22O
Molecular Weight: 170.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: 4-Pentylcyclohexanol

Formula: C₁₁H₂₂O
Density: 0.90 (20 °C)
MDL Number: MFCD01017412
CAS Number: 54410-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d40dcc13b5c2d6da27168ff12bed59c7e2e421ac&vid=d40dcc13b5c2d6da27168ff12bed59c7e2e421ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR