Nonaethylene glycol monomethyl ether ≥93.0%

Supplier: TCI America

Synonyms: 2,5,8,11,14,17,20,23,26-Nonaoxaoctacosan-28-ol, mPEG9-Alcohol

N0699-500MG N0699-1G
TCN0699-500MGEA 198.29 CAD
TCN0699-500MG TCN0699-1G
Nonaethylene glycol monomethyl ether ≥93.0%
Nonaethylene glycol monomethyl ether

CAS Number: 6048-68-6
MDL Number: MFCD06797148
Molecular Formula: C19H40O10
Molecular Weight: 428.52
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Melting point (°C): 14
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C


Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: mPEG9-Alcohol

Formula: C₁₉H₄₀O₁₀
Melting Pt: 14 °C
Density: 1.10 (20 °C)
MDL Number: MFCD06797148
CAS Number: 6048-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b00b60289937ef35c3a854930a6a762c013b76&vid=11b00b60289937ef35c3a854930a6a762c013b76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR