3-Nitrobenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

N0696-5G N0696-25G
TCN0696-5GEA 26.86 CAD
TCN0696-5G TCN0696-25G
3-Nitrobenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
3-Nitrobenzenesulphonamide

CAS Number: 121-52-8
MDL Number: MFCD00007935
Molecular Formula: C6H6N2O4S
Molecular Weight: 202.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₆H₆N₂O₄S
MDL Number: MFCD00007935
CAS Number: 121-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99d2905bd41c72831134796221cab4a23a89180f&vid=99d2905bd41c72831134796221cab4a23a89180f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR