Dimethyl-2,7-naphthalenedicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: 2,7-Naphthalenedicarboxylic Acid Dimethyl Ester

N0684-1G N0684-5G
TCN0684-1GEA 472.74 CAD
TCN0684-1G TCN0684-5G
Dimethyl-2,7-naphthalenedicarboxylate ≥98.0%
Dimethyl-2,7-naphthalenedicarboxylate

CAS Number: 2549-47-5
MDL Number: MFCD03427560
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: 2,7-Naphthalenedicarboxylic Acid Dimethyl Ester

Formula: C₁₄H₁₂O₄
MW: 244.25 g/mol
Melting Pt: 138 °C
MDL Number: MFCD03427560
CAS Number: 2549-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=100d65d19dd42194bc9252ecf0d3d049915a4520&vid=100d65d19dd42194bc9252ecf0d3d049915a4520&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR