3-(2-Naphthyl)-L-alanine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: H-2-Nal-OH·HCl, (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride

N0683-1G N0683-5G
TCN0683-1GEA 224.83 CAD
TCN0683-1G TCN0683-5G
3-(2-Naphthyl)-L-alanine hydrochloride ≥98.0%
3-(2-Naphthyl)-L-alanine hydrochloride

CAS Number: 122745-12-4
MDL Number: MFCD03095595
Molecular Formula: C13H13NO2
Molecular Weight: 251.71
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -9 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride

Formula: C₁₃H₁₃NO₂·HCl
MW: 251.71 g/mol
MDL Number: MFCD03095595
CAS Number: 122745-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5163514fdbdf42d490aa93dc837ca72aa98169a5&vid=5163514fdbdf42d490aa93dc837ca72aa98169a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR