(S)-2-Aminopropanamide hydrochloride ≥97.0%

Supplier: TCI America

Synonyms: H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride

A1919-25G A1919-5G
TCA1919-25GEA 120.08 CAD
TCA1919-25G TCA1919-5G
(S)-2-Aminopropanamide hydrochloride ≥97.0%
(S)-2-Aminopropanamide hydrochloride

CAS Number: 33208-99-0
Molecular Formula: C3H8N2O
Molecular Weight: 124.57
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 217
Specific rotation [a]20/D: 11 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acid Amides
Other Product Names / Synonyms: (S)-2-Aminopropionamide Hydrochloride

Formula: C₃H₉ClN₂O
MW: 124.57 g/mol
Melting Pt: 217 °C
MDL Number: MFCD00066145
CAS Number: 33208-99-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c19f375eff925ad4467510545c58701b5352807b&vid=c19f375eff925ad4467510545c58701b5352807b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR