Ethyl-(S)-3-piperidinecarboxylate-D-tartrate ≥98.0%

Supplier: TCI America

Synonyms: (S)-3-Piperidinecarboxylic acid ethyl ester D-Tartrate, Ethyl (S)-Nipecotate D-Tartrate, Ethyl (S)-3-Piperidinecarboxylate D-tartrate, (S)-Nipecotic acid ethyl ester D-Tartrate

N0681-5G N0681-25G
TCN0681-5GEA 162.74 CAD
TCN0681-5G TCN0681-25G
Ethyl-(S)-3-piperidinecarboxylate-D-tartrate ≥98.0%
Ethyl-(S)-3-piperidinecarboxylate-D-tartrate

CAS Number: 83602-38-4
MDL Number: MFCD03093639
Molecular Formula: C8H15NO2
Molecular Weight: 307.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 156
Specific rotation [a]20/D: -10.3 deg (C=5, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-Nipecotic Acid Ethyl Ester D-Tartrate, Ethyl (S)-Nipecotate D-Tartrate, (S)-3-Piperidinecarboxylic Acid Ethyl Ester D-Tartrate

Formula: C₁₂H₂₁NO₈
MW: 307.30 g/mol
Melting Pt: 156 °C
MDL Number: MFCD03093639
CAS Number: 163343-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=681f8d730b7b1bfb1509ecf6e73385132eaebd77&vid=681f8d730b7b1bfb1509ecf6e73385132eaebd77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR