(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate ≥98.0%

Supplier: TCI America

Synonyms: (R)-Ethyl piperidine-3-carboxylate (2S,3S)-2,3-dihydroxysuccinate, Ethyl (R)-Nipecotate L-Tartrate, (R)-Nipecotic acid ethyl ester L-Tartrate, (R)-3-Piperidinecarboxylic acid ethyl ester L-Tartrate, Ethyl (R)-nipecotate L-tartrate, Ethyl (R)-3-Piperidinecarboxylate L-Tartrate

N0680-25G N0680-5G
TCN0680-25GEA 403.37 CAD
TCN0680-25G TCN0680-5G
(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate ≥98.0%
(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate

CAS Number: 167392-57-6
MDL Number: MFCD00799538
Molecular Formula: C8H15NO2
Molecular Weight: 307.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 157
Specific rotation [a]20/D: 10.3 deg (C=5, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl (R)-Nipecotate L-Tartrate, (R)-Nipecotic Acid Ethyl Ester L-Tartrate, (R)-3-Piperidinecarboxylic Acid Ethyl Ester L-Tartrate

Formula: C₁₂H₂₁NO₈
MW: 307.30 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00799538
CAS Number: 167392-57-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc559c971b8f7d0d3d3342a8c3a72cfff7388071&vid=fc559c971b8f7d0d3d3342a8c3a72cfff7388071&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR