(S)-(+)-Ethyl-3-piperidinecarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl L-nipecotate, Ethyl (S)-(+)-3-Piperidinecarboxylate, (S)-Ethyl piperidine-3-carboxylate
CAS Number: 37675-18-6
MDL Number: MFCD00792499
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: 1 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl (S)-(+)-Nipecotate, (S)-(+)-Nipecotic Acid Ethyl Ester, (S)-(+)-3-Piperidinecarboxylic Acid Ethyl Ester
Formula:
C₈H₁₅NO₂ MW: 157.21 g/mol Density: 1.02 (20 °C) |
MDL Number:
MFCD00792499 CAS Number: 37675-18-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...