(S)-(+)-Nipecotic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0678-5G N0678-1G
TCN0678-5GEA 145.36 CAD
TCN0678-5G TCN0678-1G
(S)-(+)-Nipecotic acid ≥98.0% (by titrimetric analysis)
(S)-(+)-Nipecotic acid

CAS Number: 59045-82-8
MDL Number: MFCD01630807
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 5.5 deg (C=5, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-(+)-Nipecotic Acid

Formula: C₆H₁₁NO₂
MDL Number: MFCD01630807
CAS Number: 59045-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2845568f0faa85aa09032a56cbe4d5d9f2c7e94&vid=a2845568f0faa85aa09032a56cbe4d5d9f2c7e94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR