Monoethyl-5-nitroisophthalate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-Nitroisophthalic Acid Monoethyl Ester
N0676-25G
N0676-500G
TCN0676-25GEA
45.82
CAD
TCN0676-25G
TCN0676-500G
Monoethyl-5-nitroisophthalate ≥98.0% (by GC, titration analysis)
Monoethyl-5-nitroisophthalate
CAS Number: 22871-55-2
MDL Number: MFCD00130051
Molecular Formula: C10H9NO6
Molecular Weight: 239.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Other Product Names / Synonyms: 5-Nitroisophthalic Acid Monoethyl Ester
Formula:
C₁₀H₉NO₆ MW: 239.18 g/mol |
MDL Number:
MFCD00130051 CAS Number: 22871-55-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...