Neohesperidin dihydrochalcone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: NHDC, NCI-c60764, Neohesperidin DC, 1-(4-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,6-dihydroxyphenyl)-3-(3-hydroxy-4-methoxyphenyl)propan-1-one

N0675-25G N0675-5G
TCN0675-25GEA 705.79 CAD
TCN0675-25G TCN0675-5G
Neohesperidin dihydrochalcone ≥98.0% (by titrimetric analysis)
Neohesperidin dihydrochalcone

CAS Number: 20702-77-6
MDL Number: MFCD03840557
Molecular Formula: C28H36O15
Molecular Weight: 612.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₂₈H₃₆O₁₅
MDL Number: MFCD00017711
CAS Number: 20702-77-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a1fe698b7975f17783d8418b7fbd8836f702ca&vid=e9a219dbd3eb944bca152a1b65c41f8c52d5f9b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR