Nicotinamide-N-oxide ≥98.0%

Supplier: TCI America

Synonyms: Nicotinamide N-oxide, Pyridine-3-carboxamide N-oxide, 3-Carbamoylpyridine 1-oxide

N0669-25G N0669-5G
TCN0669-25GEA 543.84 CAD
TCN0669-25G TCN0669-5G
Nicotinamide-N-oxide ≥98.0%
Nicotinamide-N-oxide

CAS Number: 1986-81-8
MDL Number: MFCD00006202
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD00006202
CAS Number: 1986-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=935c04e9c2080218570ffe3726958ca762aa1c07&vid=935c04e9c2080218570ffe3726958ca762aa1c07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR