3-(2-Naphthyl)-D-alanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0665-1G N0665-5G
TCN0665-001GEA 106.1 CAD
TCN0665-001G TCN0665-5G
3-(2-Naphthyl)-D-alanine ≥98.0% (by HPLC, titration analysis)
3-(2-Naphthyl)-D-alanine

CAS Number: 76985-09-6
MDL Number: MFCD00038546
Molecular Formula: C13H13NO2
Molecular Weight: 215.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 13 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-alpha-Amino-2-naphthalenepropionic Acid

Formula: C₁₃H₁₃NO₂
MDL Number: MFCD00038546
CAS Number: 76985-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dab27ba433b0c22202016cacaee93becc3ecec1&vid=6dab27ba433b0c22202016cacaee93becc3ecec1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR