Nω-Nitro-L-arginine methyl ester hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, L-NAME Hydrochloride, L-NAME HCl, H-Arg(NO2)-OMe.HCl, (S)-Methyl 2-amino-5-(3-nitroguanidino)pentanoate hydrochloride
CAS Number: 51298-62-5
MDL Number: MFCD00039052
Molecular Formula: C7H15N5O4
Molecular Weight: 269.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 159
Specific rotation [a]20/D: 15 deg (C=3, MeOH)
Product Categories: Bioscience, Reagents for Life-style related Disease Models, non-Proteinogenic Amino Acids, Amino Acids (C-Protected), Reagents for Nitric Oxide Research
Other Product Names / Synonyms: H-Arg(NO2)-OMe.HCl, L-NAME Hydrochloride
Formula:
C₇H₁₅N₅O₄ MW: 269.69 g/mol Melting Pt: 159 °C |
MDL Number:
MFCD00039052 CAS Number: 51298-62-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...