Nω-Nitro-L-arginine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: L-NNA, H-Arg(NO2)-OH

N0660-25G N0660-5G
TCN0660-025GEA 213.3 CAD
TCN0660-025G TCN0660-005G
Nω-Nitro-L-arginine ≥98.0% (by HPLC, titration analysis)
Nomega-Nitro-L-arginine

CAS Number: 2149-70-4
MDL Number: MFCD00007033
Molecular Formula: C6H13N5O4
Molecular Weight: 219.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 24 deg (C=2, 2mol/L Hcl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: H-Arg(NO2)-OH, L-NNA

Formula: C₆H₁₃N₅O₄
MW: 219.20 g/mol
MDL Number: MFCD00007033
CAS Number: 2149-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f809b4e041e7fe33b932a22773350a97bb9feac0&vid=f809b4e041e7fe33b932a22773350a97bb9feac0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR