Nile red

Supplier: TCI America

Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

N0659-1G N0659-500MG N0659-5G
TCN0659-1GEA 557.11 CAD
TCN0659-1G TCN0659-500MG TCN0659-5G
Nile red
Nile red

CAS Number: 7385-67-3
MDL Number: MFCD00011639
Molecular Formula: C20H18N2O2
Molecular Weight: 318.38
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

Formula: C₂₀H₁₈N₂O₂
MW: 318.38 g/mol
MDL Number: MFCD00011639
CAS Number: 7385-67-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b3a5fca972f39f5674b359d5b66ca443a3a88c4&vid=8b3a5fca972f39f5674b359d5b66ca443a3a88c4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR