(R)-(-)-Nipecotic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0654-5G N0654-1G
TCN0654-5GEA 170.64 CAD
TCN0654-5G TCN0654-1G
(R)-(-)-Nipecotic acid ≥98.0% (by titrimetric analysis)
(R)-(-)-Nipecotic acid

CAS Number: 25137-00-2
MDL Number: MFCD01630787
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -5.5 deg (C=5, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-(-)-Nipecotic Acid

Formula: C₆H₁₁NO₂
MDL Number: MFCD01630787
CAS Number: 25137-00-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30619bf8c655c4fb71270f3a94f3ede46528afc8&vid=30619bf8c655c4fb71270f3a94f3ede46528afc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR