8-Nonen-1-ol ≥98.0%

Supplier: TCI America

N0650-5G N0650-25G
TCN0650-005GEA 292.3 CAD
TCN0650-005G TCN0650-025G
8-Nonen-1-ol ≥98.0%
8-Nonen-1-ol

CAS Number: 13038-21-6
MDL Number: MFCD00798062
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Specific Gravity (20/20): 0.85

Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 134 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00798062
CAS Number: 13038-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=539b801d35fd83ac31b2da8b8fa1004c7c23c6f9&vid=539b801d35fd83ac31b2da8b8fa1004c7c23c6f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR