3-(2-Naphthyl)-L-alanine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

N0646-5G N0646-1G
TCN0646-5GEA 818.12 CAD
TCN0646-5G TCN0646-001G
3-(2-Naphthyl)-L-alanine ≥97.0% (by titrimetric analysis)
3-(2-Naphthyl)-L-alanine

CAS Number: 58438-03-2
MDL Number: MFCD00066087
Molecular Formula: C13H13NO2
Molecular Weight: 215.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 240
Specific rotation [a]20/D: -11 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-alpha-Amino-2-naphthalenepropionic Acid

Formula: C₁₃H₁₃NO₂
Melting Pt: 240 °C
MDL Number: MFCD00066087
CAS Number: 58438-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29d3e9bfadeb7341917e3f0f1d59443fc2ad9600&vid=29d3e9bfadeb7341917e3f0f1d59443fc2ad9600&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR