5-Norbornene-2,2-dimethanol ≥98.0%

Supplier: TCI America

Synonyms: 2,2-Bis(hydroxymethyl)-5-norbornene, Bicyclo[2.2.1]hept-5-ene-2,2-dimethanol

N0636-1G N0636-5G
TCN0636-001GEA 135.88 CAD
TCN0636-001G TCN0636-5G
5-Norbornene-2,2-dimethanol ≥98.0%
5-Norbornene-2,2-dimethanol

CAS Number: 6707-12-6
MDL Number: MFCD00167595
Molecular Formula: C9H14O2
Molecular Weight: 154.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2,2-dimethanol, 2,2-Bis(hydroxymethyl)-5-norbornene

Formula: C₉H₁₄O₂
MW: 154.21 g/mol
Melting Pt: 169 °C
MDL Number: MFCD00167595
CAS Number: 6707-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f20a43bb6d76c1e80b5b9cc2c02144e378c50631&vid=f20a43bb6d76c1e80b5b9cc2c02144e378c50631&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR