Sodium-2-naphthoate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0617-5G N0617-25G
TCN0617-005GEA 140.46 CAD
TCN0617-005G TCN0617-025G
Sodium-2-naphthoate ≥98.0% (by titrimetric analysis)
Sodium-2-naphthoate

CAS Number: 17273-79-9
MDL Number: MFCD00671570
Molecular Formula: C11H8O2
Molecular Weight: 194.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Naphthalenecarboxylic Acid Sodium Salt, 2-Naphthoic Acid Sodium Salt

Formula: C₁₁H₈O₂
MDL Number: MFCD00671570
CAS Number: 17273-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbecee261531ae5668b6908272e0761cf9fe25ba&vid=cbecee261531ae5668b6908272e0761cf9fe25ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR