(4-Nitrophenyl)-β-D-galactopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
[Substrate for beta-Galactosidase]
CAS Number: 3150-24-1
MDL Number: MFCD00063256
Molecular Formula: C12H15NO8
Molecular Weight: 301.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 180
Specific rotation [a]20/D: -84 deg (C=1, H2O)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, p-Nitrophenyl Glycosides, Bioscience, Glycosides, Galactose, p-Nitrophenyl Glycosides, ƒÀ-Galactosidase Substrates
Other Product Names / Synonyms: PNPG
Formula:
C₁₂H₁₅NO₈ Melting Pt: 180 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063256 CAS Number: 3150-24-1 EINECS: 221-584-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...