2-Acetamidopyridine ≥98.0%
Supplier: TCI America
A1911-1G
A1911-5G
TCA1911-1GEA
84.37
CAD
TCA1911-1G
TCA1911-5G
2-Acetamidopyridine ≥98.0%
2-Acetamidopyridine
CAS Number: 5231-96-9
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 69
Formula:
C₇H₈N₂O MW: 136.15 g/mol Boiling Pt: 148 °C Melting Pt: 69 °C |
MDL Number:
MFCD00023423 CAS Number: 5231-96-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...