5,12-Naphthacenequinone ≥98.0%
Supplier: TCI America
N0603-10G
N0603-25G
TCN0603-010GEA
450.3
CAD
TCN0603-010G
TCN0603-25G
5,12-Naphthacenequinone ≥98.0%
5,12-Naphthacenequinone
CAS Number: 1090-13-7
MDL Number: MFCD00003701
Molecular Formula: C18H10O2
Molecular Weight: 258.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 2,3-Benzo-9,10-anthraquinone, Tetracenequinone
Formula:
C₁₈H₁₀O₂ |
MDL Number:
MFCD00003701 CAS Number: 1090-13-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...