3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol ≥97.0% for synthesis

Supplier: TCI America

Synonyms: 1H,1H,2H,2H-Perfluoro-1-hexanol, 1H,1H,2H,2H-Nonafluoro-1-hexanol, 1H,1H,2H,2H-Perfluorohexan-1-ol

N0600-25G N0600-5G
TCN0600-025GEA 169.06 CAD
TCN0600-025G TCN0600-5G
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol ≥97.0% for synthesis
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol

CAS Number: 2043-47-2
MDL Number: MFCD00039543
Molecular Formula: C6H5F9O
Molecular Weight: 264.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 73
Specific Gravity (20/20): 1.59


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 2-(Perfluorobutyl)ethanol

Formula: C₆H₅F₉O
Boiling Pt: 76 °C
Density: 1.59 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00039543
CAS Number: 2043-47-2
EINECS: 218-050-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0e3ead6b7a62b53c528c84d82516ca6fea086d2&vid=f0e3ead6b7a62b53c528c84d82516ca6fea086d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR