Dimethyl-2,3-naphthalenedicarboxylate ≥97.0%
Supplier: TCI America
Synonyms:
2,3-Naphthalenedicarboxylic Acid Dimethyl Ester
N0590-25G
N0590-1G
N0590-5G
TCN0590-25GEA
844.67
CAD
TCN0590-25G
TCN0590-001G
TCN0590-5G
Dimethyl-2,3-naphthalenedicarboxylate ≥97.0%
Dimethyl-2,3-naphthalenedicarboxylate
CAS Number: 13728-34-2
MDL Number: MFCD00012278
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 50
Other Product Names / Synonyms: 2,3-Naphthalenedicarboxylic Acid Dimethyl Ester
Formula:
C₁₄H₁₂O₄ Boiling Pt: 145 °C Melting Pt: 50 °C |
MDL Number:
MFCD00012278 CAS Number: 13728-34-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...