4-Nitropicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0589-1G N0589-100MG
TCN0589-1GEA 634.69 CAD
TCN0589-1G TCN0589-100MG
4-Nitropicolinic acid ≥98.0% (by GC, titration analysis)
4-Nitropicolinic acid

CAS Number: 13509-19-8
MDL Number: MFCD00191562
Molecular Formula: C6H4N2O4
Molecular Weight: 168.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Nitropicolinic Acid

Formula: C₆H₄N₂O₄
MDL Number: MFCD00191562
CAS Number: 13509-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1abc6d5ad402985a55f8cc4b8997e2bff9ce19ab&vid=1abc6d5ad402985a55f8cc4b8997e2bff9ce19ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR