3-(5-Nitro-2-furyl)acrylic acid ≥97.0% (by HPLC)
Supplier: TCI America
N0551-5G
N0551-25G
TCN0551-005GEA
235.42
CAD
TCN0551-005G
TCN0551-25G
3-(5-Nitro-2-furyl)acrylic acid ≥97.0% (by HPLC)
3-(5-Nitro-2-furyl)acrylic acid
CAS Number: 6281-23-8
MDL Number: MFCD00143078
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Formula:
C₇H₅NO₅ |
MDL Number:
MFCD00143078 CAS Number: 6281-23-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...