trans-4-Nitrostilbene ≥98.0%

Supplier: TCI America

N0549-25G N0549-5G
TCN0549-025GEA 311.26 CAD
TCN0549-025G TCN0549-5G
trans-4-Nitrostilbene ≥98.0%
trans-4-Nitrostilbene

CAS Number: 1694-20-8
MDL Number: MFCD00053614
Molecular Formula: C14H11NO2
Molecular Weight: 225.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 157

Formula: C₁₄H₁₁NO₂
Melting Pt: 157 °C
MDL Number: MFCD00053614
CAS Number: 1694-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fea72410a18fa0182ccd283f5d74a262f0b44302&vid=0f07543e955290242ba85e25e639df99cb35f14b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR