1,4,5,8-Naphthalenetetracarboxdiimide ≥97.0% (by total nitrogen basis)

Supplier: TCI America

N0536-1G N0536-5G
TCN0536-001GEA 303.36 CAD
TCN0536-001G TCN0536-5G
1,4,5,8-Naphthalenetetracarboxdiimide ≥97.0% (by total nitrogen basis)
1,4,5,8-Naphthalenetetracarboxdiimide

CAS Number: 5690-24-4
MDL Number: MFCD00047649
Molecular Formula: C14H6N2O4
Molecular Weight: 266.21
Purity/Analysis Method: >97.0% (N)
Form: Crystal

Formula: C₁₄H₆N₂O₄
MDL Number: MFCD00047649
CAS Number: 5690-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b9311b172f78170f6bc14d491768b95e211779&vid=c2b9311b172f78170f6bc14d491768b95e211779&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR