3-Nonylthiophene
Supplier: TCI America
N0533-1G
N0533-5G
TCN0533-001GEA
148.84
CAD
TCN0533-001G
TCN0533-005G
3-Nonylthiophene
3-Nonylthiophene
CAS Number: 65016-63-9
MDL Number: MFCD00130137
Molecular Formula: C13H22S
Molecular Weight: 210.38
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.92
Formula:
C₁₃H₂₂S Density: 0.92 (20 °C) |
MDL Number:
MFCD00130137 CAS Number: 65016-63-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...