2,3-Naphthalenedicarboxylic anhydride ≥95.0% (by GC, titration analysis)

Supplier: TCI America

N0530-25G N0530-250G
TCN0530-025GEA 336.86 CAD
TCN0530-025G TCN0530-250G
2,3-Naphthalenedicarboxylic anhydride ≥95.0% (by GC, titration analysis)
2,3-Naphthalenedicarboxylic anhydride

CAS Number: 716-39-2
MDL Number: MFCD00059091
Molecular Formula: C12H6O3
Molecular Weight: 198.18
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Boiling point (°C): 380
Melting point (°C): 248

Formula: C₁₂H₆O₃
Boiling Pt: 380 °C
Melting Pt: 248 °C
MDL Number: MFCD00059091
CAS Number: 716-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54d50b5d3c0af281ecc8b656b3ba12872a77096&vid=f54d50b5d3c0af281ecc8b656b3ba12872a77096&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR