2-Nitro-p-xylylene glycol ≥95.0%

Supplier: TCI America

Synonyms: (2-Nitro-1,4-phenylene)dimethanol

N0519-5G N0519-1G
TCN0519-005GEA 446.19 CAD
TCN0519-005G TCN0519-1G
2-Nitro-p-xylylene glycol ≥95.0%
2-Nitro-p-xylylene glycol

CAS Number: 23222-97-1
MDL Number: MFCD00024286
Molecular Formula: C8H9NO4
Molecular Weight: 183.16
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 95

Formula: C₈H₉NO₄
MW: 183.16 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00024286
CAS Number: 23222-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355303c2904c22536aa0b0e03c5ab7435f131f80&vid=355303c2904c22536aa0b0e03c5ab7435f131f80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR