4-Nitropyrrole-2-carboxylic acid hydrate ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
N0509-5G
N0509-1G
TCN0509-005GEA
474
CAD
TCN0509-005G
TCN0509-001G
4-Nitropyrrole-2-carboxylic acid hydrate ≥99.0% (by HPLC, titration analysis)
4-Nitropyrrole-2-carboxylic acid hydrate
CAS Number: 5930-93-8
MDL Number: MFCD00059928
Molecular Formula: C5H4N2O4
Molecular Weight: 156.10
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 217
Formula:
C₅H₄N₂O₄ Melting Pt: 217 °C |
MDL Number:
MFCD00059928 CAS Number: 5930-93-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...