3-Nitropyrrole ≥98.0%

Supplier: TCI America

N0502-5G N0502-1G
TCN0502-5GEA 1168.57 CAD
TCN0502-5G TCN0502-001G
3-Nitropyrrole ≥98.0%
3-Nitropyrrole

CAS Number: 5930-94-9
MDL Number: MFCD00059714
Molecular Formula: C4H4N2O2
Molecular Weight: 112.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 100

Formula: C₄H₄N₂O₂
Melting Pt: 100 °C
MDL Number: MFCD00059714
CAS Number: 5930-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=305980abbfccfb975ffa8f7779308ed3b0cb2896&vid=305980abbfccfb975ffa8f7779308ed3b0cb2896&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR