1-Aminohydantoin hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
1-Aminohydantoin Hydrochloride, 1-Aminohydantoin HCl, 1-Amino-2,4-imidazolidinedione hydrochloride
A1902-25G
A1902-5G
TCA1902-25GEA
265.44
CAD
TCA1902-25G
TCA1902-5G
1-Aminohydantoin hydrochloride ≥98.0%
1-Aminohydantoin hydrochloride
CAS Number: 2827-56-7
Molecular Formula: C3H5N3O2
Molecular Weight: 151.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 203
Formula:
C₃H₅N₃O₂ MW: 151.55 g/mol Melting Pt: 203 °C |
MDL Number:
MFCD02181047 CAS Number: 2827-56-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...