Nonadecanoic acid ethyl ester ≥96.0%
Supplier: TCI America
N0459-25G
N0459-5G
TCN0459-025GEA
337.17
CAD
TCN0459-025G
TCN0459-5G
Nonadecanoic acid ethyl ester ≥96.0%
Nonadecanoic acid ethyl ester
CAS Number: 18281-04-4
MDL Number: MFCD00026690
Molecular Formula: C21H42O2
Molecular Weight: 326.57
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Freezing point (°C): 35
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Nonadecanoic Acid Ethyl Ester
Formula:
C₂₁H₄₂O₂ |
MDL Number:
MFCD00026690 CAS Number: 18281-04-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...