Neopentyl glycol diglycidyl Ether 40,0%

Supplier: TCI America

Synonyms: Diglycidyloxyneopentane, Neopentyl glycol diglycidyl ether

N0448-25ML N0448-500ML
TCN0448-025MLEA 53.02 CAD
TCN0448-025ML TCN0448-500ML
Neopentyl glycol diglycidyl Ether 40,0%
Neopentyl glycol diglycidyl Ether

CAS Number: 17557-23-2
MDL Number: MFCD00040687
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method: >40.0% (GC)
Form: Clear Liquid
Flash Point (°C): 141
Specific Gravity (20/20): 1.08


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Diglycidyloxyneopentane

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Density: 1.08 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00040687
CAS Number: 17557-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=689e7968001230e84686ce5f075621381c4c2528&vid=689e7968001230e84686ce5f075621381c4c2528&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 40.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR