6-Nitrofluorescein (isomer II) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3',6'-Dihydroxy-6-nitro-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 6-Nitro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one
N0445-5G
N0445-1G
TCN0445-005GEA
1077.56
CAD
TCN0445-005G
TCN0445-001G
6-Nitrofluorescein (isomer II) ≥98.0% (by titrimetric analysis)
6-Nitrofluorescein (isomer II)
CAS Number: 27402-68-2
MDL Number: MFCD00059694
Molecular Formula: C20H11NO7
Molecular Weight: 377.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 6-Nitro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one
Formula:
C₂₀H₁₁NO₇ MW: 377.31 g/mol |
MDL Number:
MFCD00059694 CAS Number: 27402-68-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...