Disodium naphthalene-1,5-disulfonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Disodium 1,5-Naphthalenedisulphonate hydrate, Naphthalene-1,5-disulphonic acid disodium salt, Naphthalene-1,5-disulfonic acid disodium salt, 1,5-Naphthalenedisulfonic acid disodium salt, 1,5-Naphthalenedisulphonic acid disodium salt, Sodium 1,5-naphthalenedisulfonate dibasic, Disodium 1,5-naphthalenedisulphonate, Disodium 1,5-naphthalenedisulfonate, Disodium 1,5-Naphthalenedisulfonate hydrate, Sodium 1,5-naphthalenedisulphonate dibasic
CAS Number: 1655-29-4
MDL Number: MFCD00064178
Molecular Formula: C10H8O6S2
Molecular Weight: 332.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 1,5-Naphthalenedisulfonic Acid Disodium Salt
Formula:
C₁₀H₆Na₂O₆S₂ MW: 332.27 g/mol |
MDL Number:
MFCD00064178 CAS Number: 1655-29-4 EINECS: 216-732-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...