2-Isobutoxynaphthalene ≥98.0%

Supplier: TCI America

Synonyms: Isobutyl 2-Naphthyl Ether

N0433-500G N0433-25G
TCN0433-500GEA 365.3 CAD
TCN0433-500G TCN0433-025G
2-Isobutoxynaphthalene ≥98.0%
2-Isobutoxynaphthalene

CAS Number: 2173-57-1
MDL Number: MFCD00046467
Molecular Formula: C14H16O
Molecular Weight: 200.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 68
Freezing point (°C): 32


Other Product Names / Synonyms: Isobutyl 2-Naphthyl Ether

Formula: C₁₄H₁₆O
MW: 200.28 g/mol
Flash Pt: 68 °C
MDL Number: MFCD00046467
CAS Number: 2173-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1bef141735d336412418370b7a7b453f281c3c2&vid=d1bef141735d336412418370b7a7b453f281c3c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR