N-(2-Carboxyethyl)iminodiacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(2-carboxyethyl)iminodiaceticacid, Nitrilodiacetic-propionic Acid

N0408-25G N0408-5G
TCN0408-025GEA 306.93 CAD
TCN0408-025G TCN0408-5G
N-(2-Carboxyethyl)iminodiacetic acid ≥98.0% (by titrimetric analysis)
N-(2-Carboxyethyl)iminodiacetic acid

CAS Number: 6245-75-6
MDL Number: MFCD00059634
Molecular Formula: C7H11NO6
Molecular Weight: 205.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 195


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Nitrilodiacetic-propionic Acid

Formula: C₇H₁₁NO₆
MW: 205.17 g/mol
Melting Pt: 195 °C
MDL Number: MFCD00059634
CAS Number: 6245-75-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3fcece48aa88396d89378f4bcb7b74c147ef770&vid=f3fcece48aa88396d89378f4bcb7b74c147ef770&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR